Anti-LCE1B Rabbit Polyclonal Antibody

Supplier: Proteintech
21771-1-AP-150UL
10089-552EA 402 USD
10089-552
Anti-LCE1B Rabbit Polyclonal Antibody
Antibodies
Human LCE1B (Late cornified envelope protein 1B), also named late envelope protein 2 (LEP2), is located at human Chromosome1q21 which contains multiple LCE clusters. LCE1B gene is predicated to encode a 118-aa protein, and is currently evidenced at transcript level. Its expression was skin-specific and was readily detected in adult trunk skin, adult arm skin, fetal skin, penal skin, vulva, esophagus and tongue . LCE genes respond to environmental stimuli such as calcium levels and ultraviolet (UV) light, however, LCE groups have distinct functions .

Western Blot:A375 Cells, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200

Type: Primary
Antigen: LCE1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR