Anti-FAM117B Rabbit Polyclonal Antibody

Supplier: Proteintech
21768-1-AP-150UL
10086-780EA 402 USD
10086-780
Anti-FAM117B Rabbit Polyclonal Antibody
Antibodies
FAM117B, also named as ALS2CR13, is mapped in the genomic region covering the complete candidate region for Amyotrophic lateral sclerosis 2 (ALS2). This antibody is specific to FAM117B.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: FAM117B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR