Anti-PDE4C Rabbit Polyclonal Antibody

Supplier: Proteintech
21754-1-AP-150UL
10092-002EA 402 USD
10092-002
Anti-PDE4C Rabbit Polyclonal Antibody
Antibodies
PDE4C(cAMP-specific 3',5'-cyclic phosphodiesterase 4C) is also named as DPDE1 and belongs to the PDE4 subfamily, which play a key role in lung inflammation and hyperresponsiveness. PDE4C promotes the hydrolysis of cAMP, and PKD2, functioningas a Ca2+ entry channel, may mediate local accumulation of Ca2+ that inhibits the activity of Ca2+-sensitive ADCY5. The three isoforms(84,75, and 64 kDa) of PDE4C have been detected in cytosolic, microsomal, and nuclear. This antibody is specific to PDE4C.

Type: Primary
Antigen: PDE4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR