Anti-HIGD1A Rabbit Polyclonal Antibody

Supplier: Proteintech
21749-1-AP-150UL
10088-248EA 402 USD
10088-248
Anti-HIGD1A Rabbit Polyclonal Antibody
Antibodies
HIG1 domain family member 1A (HIGD1A) is a proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. May play a role in the assembly of respiratory supercomplexes (22342701). It is induced by hypoxia induction.This antibody is a rabbit polyclonal antibody raised against a full-length human HIGD1A protein.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: HIGD1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR