Anti-VNN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21745-1-AP-150UL
10096-994EA 524.48 USD
10096-994
Anti-VNN1 Rabbit Polyclonal Antibody
Antibodies
VNN1(Vascular non-inflammatory molecule 1) is also named as Vanin-1,Pantetheinase,Tiff66.It is a GPI-anchored 70-kDa protein with a high degree of homology with GPI-80 (also known as VNN2), a molecule expressed by human phagocytes and involved in leukocyte adhesion and migration.Vanin-1 is predominantly expressed by resident tissue cells and acts at the level of epithelial cells and surrounding hematopoietic cells by paracrine cysteamine release. The full length VNN1 has a signal peptide, a propeptide and six glycosylation sites. According to some authors, the variations which varied between 55 and 72kDa may be due to different degrees of glycosylation of the protein and cleavage(JOSE A et al. 2001).

Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: VNN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR