Anti-CYP2A6 Rabbit Polyclonal Antibody

Supplier: Proteintech
21721-1-AP-150UL
10085-514EA 402 USD
10085-514
Anti-CYP2A6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: CYP2A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR