Anti-HLA-DRB5 Rabbit Polyclonal Antibody

Supplier: Proteintech
21702-1-AP-150UL
10088-308EA 402 USD
10088-308
Anti-HLA-DRB5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: HLA-DRB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR