Anti-DCLK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21699-1-AP-150UL
10085-698EA 402 USD
10085-698
Anti-DCLK1 Rabbit Polyclonal Antibody
Antibodies
DCLK1(Serine/threonine-protein kinase DCLK1) is also named as DCAMKL1, DCDC3A, KIAA0369 and belongs to the CAMK Ser/Thr protein kinase family. It is a microtubule-associated kinase that can undergo autophosphorylation and it also has microtubule-polymerizing activity that is independent of its protein kinase activity. It plays a unique role in mitotic spindle integrity during early neurogenesis in radial glial cell proliferation and their radial process stability. DCLK1 is a unique marker for distinguishing tumor stem cells from intestinal normal stem cells. This protein has 4 isoforms produced by alternative splicing with the molecular weight of 82 kDa, 81 kDa,47 kDa and 48 kDa.

Western Blot: Human Brain Tissue, 1:1000-1:10000

Type: Primary
Antigen: DCLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR