Anti-NEDD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21698-1-AP-150UL
10091-008EA 537.7 USD
10091-008
Anti-NEDD4 Rabbit Polyclonal Antibody
Antibodies
NEDD4-1(Neural precursor cell expressed developmentally down-regulated protein 4) is also named as NEDD4 and KIAA0093. It plays an essential role for the activation of the genetic program involved in the terminal maturation stage of invariant natural killer T-cell development. And it is differentially expressed in mammary epithelial cells and plays a role in luminal lineage determination. NEDD4-1 is a key regulator of FGFR1 endocytosis and signalling during neuronal differentiation and embryonic development. This antibody can recognize NEDD4-1 and NEDD4-2.

Western Blot:C6 Cells, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: NEDD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR