Anti-ZNF132 Rabbit Polyclonal Antibody

Supplier: Proteintech
21681-1-AP-150UL
10097-300EA 402 USD
10097-300
Anti-ZNF132 Rabbit Polyclonal Antibody
Antibodies
ZNF132 also named as Zinc finger protein 132 is a 706 amino-acid protein, which contains 18 C2H2-type zinc fingers and 1 KRAB domain. ZNF132 belongs to the krueppel C2H2-type zinc-finger protein family and localizes in the nucleus. ZNF132 may be involved in transcriptional regulation. ZNF132 exists as two alternatively spliced isoforms, and is encoded by a gene that maps to human chromosome 19.

Western Blot: Human Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: ZNF132
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR