Anti-CNGA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21657-1-AP-150UL
10084-918EA 402 USD
10084-918
Anti-CNGA3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: CNGA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR