Anti-ZNF192 Rabbit Polyclonal Antibody

Supplier: Proteintech
21656-1-AP-150UL
10097-312EA 402 USD
10097-312
Anti-ZNF192 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: ZNF192
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR