Anti-SLITRK3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21649-1-AP-150UL
10094-818EA 402 USD
10094-818
Anti-SLITRK3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: SLITRK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR