Anti-KIF5B Rabbit Polyclonal Antibody

Supplier: Proteintech
21632-1-AP-150UL
10089-242EA 402 USD
10089-242
Anti-KIF5B Rabbit Polyclonal Antibody
Antibodies
Western Blot: Multi-Cells, 1:200-1:1000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: KIF5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR