Anti-TMED9 Rabbit Polyclonal Antibody

Supplier: Proteintech
21620-1-AP-150UL
10096-070EA 527.45 USD
10096-070
Anti-TMED9 Rabbit Polyclonal Antibody
Antibodies
TMED9 (also named as GMP25, GP25L2 or p25) is a member of the EMP24/GP25L family. TMED9 probably forms hetero-oligomeric complexes with TMED7 (p27), TMED2 (p24), and TMED10 (p23) . Located in the endoplasmic reticulum and the Golgi, TMED9 appears to be involved in vesicular protein trafficking, mainly in the early secretory pathway .

Western Blot: Mouse Liver Tissue, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: TMED9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR