Anti-FHL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21619-1-AP-150UL
10087-006EA 402 USD
10087-006
Anti-FHL2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse ovary Tissue, 1:1000-1:10000; IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: FHL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR