Anti-CDK14 Rabbit Polyclonal Antibody

Supplier: Proteintech
21612-1-AP-150UL
10092-130EA 524.34 USD
10092-130
Anti-CDK14 Rabbit Polyclonal Antibody
Antibodies
PFTK1 is a Cdc2-related protein kinase, also named as PFTAIRE1, CDK14 and belongs to the CMGC Ser/Thr protein kinase family and CDC2/CDKX subfamily. It acts as a cyclin-dependent kinases that regulates cell cycle progression and cell proliferation. The PFTK1 protein shows high homology with mouse Pftaire1, a Cdc2-related protein expressed primarily in the postnatal and adult nervous system. The protein contains 2 predicted nuclear localization signals that the N-terminal nuclear localization signals do not play a role in vivo. It has 3 isoforms produced by alternative splicing. This antibody is specific to PFTK1.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: CDK14
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR