Anti-MAP3K4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21610-1-AP-150UL
10089-938EA 402 USD
10089-938
Anti-MAP3K4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:1000-1:10000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: MAP3K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR