Anti-DLL4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21584-1-AP-150UL
10085-906EA 402 USD
10085-906
Anti-DLL4 Rabbit Polyclonal Antibody
Antibodies
DLL4, also named as Delta4, plays a role in the Notch signaling pathway. It activates Notch-1 and Notch-4. Notch signaling is activated upon engagement of the Notch receptor with its ligands, the DSL (Delta, Serrate, Lag2) proteins of single-pass type I membrane proteins. DLL4 is highly expressed in the vascular endothelium.

Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: DLL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR