Anti-CYP24A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21582-1-AP-150UL
10085-508EA 402 USD
10085-508
Anti-CYP24A1 Rabbit Polyclonal Antibody
Antibodies
CYP24A1(1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial) catalyzes the NADPH-dependent 24-hydroxylation of calcidiol (25-hydroxyvitamin D3) and calcitriol (1-alpha,25-dihydroxyvitamin D3).Cholecalciferol is metabolised by the 25-hydroxylases (CYP2R1 and CYP27A1) in the liver, before 25-hydroxycholecalciferol (25(OH)D3) relocates to the circulation.It It has two isforms with the molecular weight 51 KDa and 59KDa,respectively.This antibody is specific to CYP24A1.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: CYP24A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR