Anti-INMT Rabbit Polyclonal Antibody

Supplier: Proteintech
21578-1-AP-150UL
10088-910EA 402 USD
10088-910
Anti-INMT Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse pancreas Tissue, 1:200-1:2000

Type: Primary
Antigen: INMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR