Anti-ZEB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21544-1-AP-150UL
10097-264EA 524.48 USD
10097-264
Anti-ZEB1 Rabbit Polyclonal Antibody
Antibodies
ZEB1(zinc-finger-enhancer binding protein 1), which is a members of the Zincfinger-Homeobox (ZFH)-family of repressors, acts as a transcriptional repressor that inhibits interleukin-2 (IL-2) gene expression. Depending on the quantity of cDNA and on the cell type, ZEB1 can enhance or repress the promoter activity of the ATP1A1 gene . Represses E-cadherin promoter and induces an epithelial-mesenchymal transition (EMT) by recruiting SMARCA4/BRG1. Represses BCL6 transcription in the presence of the corepressor CTBP1. Positively regulates neuronal differentiation. This is a rabbit polyclonal antibody raised against part of the full length of ZEB1 of human.

Western Blot: MCF7 Cells, 1:500-1:5000; IP: MCF-7 Cells, 1:200-1:2000

Type: Primary
Antigen: ZEB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR