Anti-FOXO4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21535-1-AP-150UL
10081-788EA 402 USD
10081-788
Anti-FOXO4 Rabbit Polyclonal Antibody
Antibodies
FOXO4, also named as AFX, AFX1 and MLLT7, is a transcription factor which involved in the regulation of the insulin signaling pathway. It binds to insulin-response elements (IREs) and can activate transcription of IGFBP1.FOXO down-regulates expression of HIF1A and suppresses hypoxia-induced transcriptional activation of HIF1A-modulated genes. It is involved in negative regulation of the cell cycle. A chromosomal aberration involving FOXO4 is found in acute leukemias. The antiboay can recognize isoform 1 and isoform Zeta of FOXO4.

Western Blot: Mouse Heart Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: FOXO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR