Anti-MYLK3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21527-1-AP-150UL
10090-730EA 537.7 USD
10090-730
Anti-MYLK3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:1000-1:10000; IHC: Human Colon, 1:20-1:200

Type: Primary
Antigen: MYLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR