Anti-EXOG Rabbit Polyclonal Antibody

Supplier: Proteintech
21523-1-AP-150UL
10086-708EA 402 USD
10086-708
Anti-EXOG Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse cerebellum Tissue, 1:500-1:5000

Type: Primary
Antigen: EXOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR