Anti-TNFRSF13B Rabbit Polyclonal Antibody

Supplier: Proteintech
21520-1-AP-150UL
10096-134EA 402 USD
10096-134
Anti-TNFRSF13B Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:200-1:1000

Type: Primary
Antigen: TNFRSF13B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR