Anti-ACTR5 Rabbit Polyclonal Antibody

Supplier: Proteintech
21505-1-AP-150UL
10083-138EA 402 USD
10083-138
Anti-ACTR5 Rabbit Polyclonal Antibody
Antibodies
Arp5 is a member of nuclear actin-related proteins (ARPs) that function as components of the chromatin- remodeling complex. Recently study reported that Arp5 regulates smooth muscle cell differentiation through interaction with myocardin (Myocd). Arp5 has been identified as negative regulator of Myocd, suppressing Myocd activity in the nucleus through tightly binding with Myocd RPEL motifs. Alternative splicing results in two variants of Arp5: a broadly expressed arp5-common, and a smooth muscle restricted arp5-sm. Expression of Arp5-sm protein is strongly suppressed in well-differentiated smooth muscle cells by partial messenger RNA decay and translational suppression. (24567363)

Western Blot: Mouse cerebellum Tissue, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: ACTR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR