Anti-CHIA Rabbit Polyclonal Antibody

Supplier: Proteintech
21484-1-AP-150UL
10084-704EA 402 USD
10084-704
Anti-CHIA Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:200-1:2000; IHC: Human Stomach, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: CHIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR