Anti-NF-H Rabbit Polyclonal Antibody

Supplier: Proteintech
21471-1-AP-150UL
10081-672EA 402 USD
10081-672
Anti-NF-H Rabbit Polyclonal Antibody
Antibodies
NEFH, also named as KIAA0845 and NFH, Belongs to the intermediate filament family. It has an important function in mature axons that is not subserved by the two smaller NF proteins. Neurofilaments are the 10nm intermediate filaments found specifically in neurons. They are a major component of the cell's cytoskeleton, and provide support for normal axonal radial growth. Neurofilaments usually contain three intermediate filament proteins: L, M, and H which are involved in the maintenance of neuronal caliber. The names given to the three major neurofilament subunits are based upon the apparent molecular weight of the mammalian subunits on SDS-PAGE:NF-L, 65-68kd; NF-M,145-160kd and NF-H, 200-220kd. This antibody recognize NEFH only.

Western Blot: Mouse Brain Tissue, 1:1000-1:10000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: NF-H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR