Anti-C20ORF30 Rabbit Polyclonal Antibody

Supplier: Proteintech
21466-1-AP-150UL
10083-834EA 402 USD
10083-834
Anti-C20ORF30 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:1000-1:10000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: C20ORF30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR