Anti-C1orf77 Rabbit Polyclonal Antibody

Supplier: Proteintech
21456-1-AP-150UL
10083-810EA 402 USD
10083-810
Anti-C1orf77 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse spleen Tissue, 1:200-1:2000

Type: Primary
Antigen: C1orf77
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR