Anti-INPP5J Rabbit Polyclonal Antibody

Supplier: Proteintech
21417-1-AP-150UL
10088-924EA 402 USD
10088-924
Anti-INPP5J Rabbit Polyclonal Antibody
Antibodies
INPP5J(Phosphatidylinositol 4,5-bisphosphate 5-phosphatase A) is also named as PIB5PA, PIPP and belongs to the inositol 1,4,5-trisphosphate 5-phosphatase type II family. INPP5J is a key signalling molecule that regulates dendritic outgrowth through activation of small GTPase signalling via interaction between FRMPD4 and ARHGEF7. It can control the resting membrane potential through a specific ion-channel-receptor-ligand interaction that brings about a large conformational change, analogous to neurotransmitter activation of ion channels at synapses. . This protein has 3 isoforms produced by alternative splicing.

Western Blot: Human Heart Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: INPP5J
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR