Anti-MYH11 Rabbit Polyclonal Antibody

Supplier: Proteintech
21404-1-AP-150UL
10081-648EA 402 USD
10081-648
Anti-MYH11 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Uterus Tissue, 1:1000-1:10000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: MYH11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR