Anti-UHRF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21402-1-AP-150UL
10096-766EA 402 USD
10096-766
Anti-UHRF1 Rabbit Polyclonal Antibody
Antibodies
UHRF1, also named as ICBP90, NP95 and RNF106, is a putative E3 ubiquitin-protein ligase. It may participate in methylation-dependent transcriptional regulation. UHRF1 binds to inverted 5'-CCAAT-3' box 2 in the TOP2A promoter, and activates TOP2A expression. It is important for G1/S transition and may be involved in DNA repair and chromosomal stability. UHRF1's ability to repress its direct target gene expression is dependent on PHD(UHRF1) binding to unmodified H3R2.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: UHRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR