Anti-U2SURP Rabbit Polyclonal Antibody

Supplier: Proteintech
21399-1-AP-150UL
10095-146EA 402 USD
10095-146
Anti-U2SURP Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Heart Tissue, 1:1000-1:10000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HEK-293 Cells, 1:10-1:100

Type: Primary
Antigen: U2SURP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR