Anti-KIAA1602 Rabbit Polyclonal Antibody

Supplier: Proteintech
21392-1-AP-150UL
10089-210EA 402 USD
10089-210
Anti-KIAA1602 Rabbit Polyclonal Antibody
Antibodies
KIAA1602, also named as NCKAP5L, has 4 isoforms. The target band is about 139-145kd for phosphorylation.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human skin, 1:20-1:200

Type: Primary
Antigen: KIAA1602
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR