Anti-CCDC153 Rabbit Polyclonal Antibody

Supplier: Proteintech
21390-1-AP-150UL
10084-170EA 402 USD
10084-170
Anti-CCDC153 Rabbit Polyclonal Antibody
Antibodies
The 45-48kd band maybe Dimer of CCDC153.

Western Blot:COLO 320 Cells, 1:500-1:5000; IHC: Human prostate hyperplasia Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CCDC153
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR