Anti-Pan-PAX Rabbit Polyclonal Antibody

Supplier: Proteintech
21383-1-AP-150UL
10091-788EA 402 USD
10091-788
Anti-Pan-PAX Rabbit Polyclonal Antibody
Antibodies
PAX genes encode a family of developmentally regulated transcription factors that have been implicated in a number of human and murine congenital disorders, as well as in tumorigenesis. These genes are defined by the presence of an evolutionarily conserved DNA binding domain, termed the paired domain. Many paired domain-containing proteins recognize a common sequence motif. The immunogen of this atnibody is the Paired box domain of PAX family. This antibody is a Pan-PAX antibody.

Western Blot:SKOV-3 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IP:SKOV-3 Cells, 1:200-1:2000

Type: Primary
Antigen: Pan-PAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR