Anti-SASS6 Rabbit Polyclonal Antibody

Supplier: Proteintech
21377-1-AP-150UL
10094-150EA 402 USD
10094-150
Anti-SASS6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: SASS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR