Anti-TMX4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21348-1-AP-150UL
10096-114EA 402 USD
10096-114
Anti-TMX4 Rabbit Polyclonal Antibody
Antibodies
TMX4(Thioredoxin-related transmembrane protein 4) is also named as KIAA1162, TXNDC13. It is a type I transmembrane protein, with a Trx-like domain facing the ER lumen and may be to enable protein folding in cooperation with these proteins consisting of folding complex in the ER. It could work as a reductase in the environment of the ER. The full length protein has a 23 amino acid signal peptide. This antibody is specific to TMX4.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: TMX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR