Anti-SESN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21346-1-AP-150UL
10093-578EA 402 USD
10093-578
Anti-SESN2 Rabbit Polyclonal Antibody
Antibodies
Sestrins, including sestrin-1 (PA26), sestrin-2 (Hi95), and sestrin-3, are 48 to 65 kDa cystein sulfinyl reductases and they modulate peroxide signaling and antioxidant defense. These proteins selectively reduce or repair hyperoxidized forms of typical 2-Cys peroxiredoxins within eukaryotes. Expression of these proteins is regulated by p53, a tumor suppressor protein. Sestrin 2 is implicated in the regulation of cell growth and survival, in cellular responses to different stress conditions, and also acts as a positive regulator of autophagy. Sestrin 2 is approximately a 57.6kDa protein (480 amino acids).

Western Blot: Mouse Thymus Tissue, 1:1000-1:10000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: SESN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR