Anti-RBM39 Rabbit Polyclonal Antibody

Supplier: Proteintech
21339-1-AP-150UL
10093-414EA 402 USD
10093-414
Anti-RBM39 Rabbit Polyclonal Antibody
Antibodies
RBM39, also named as HCC1 or RNPC2, is a 530 amino acid protein, which contains three RRM (RNA recognition motif) domains and belongs to the splicing factor SR family. RBM39 is widely expressed in various tissues and with highly expression in pancreas, skeletal muscle, lung and brain. RBM39 is a transcriptional coactivator for steroid nuclear receptors ESR1/ER-alpha and ESR2/ER-beta, and JUN/AP-1. RBM39 may be involved in pre-mRNA splicing process.

Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: RBM39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR