Anti-C3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21337-1-AP-150UL
10083-850EA 402 USD
10083-850
Anti-C3 Rabbit Polyclonal Antibody
Antibodies
The complement system is an important effector which bridges the innate and adaptive immune systems. The third component of complement, C3, plays a central role in the activation of the complement system. Its processing by C3 convertase is the central reaction in both classical and alternative complement pathways.

  • Western Blot: Human Blood Tissue, 1:500-1:5000

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR