Anti-FAM129A Rabbit Polyclonal Antibody

Supplier: Proteintech
21333-1-AP-150UL
10086-794EA 402 USD
10086-794
Anti-FAM129A Rabbit Polyclonal Antibody
Antibodies
Western Blot: HT-1080 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IHC: Human thyroid cancer Tissue, 1:20-1:200

Type: Primary
Antigen: FAM129A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR