Anti-GTSE1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21319-1-AP-150UL
10087-986EA 402 USD
10087-986
Anti-GTSE1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:500-1:5000; IHC: Human breast cancer, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: GTSE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR