Anti-DEPDC6 Rabbit Polyclonal Antibody

Supplier: Proteintech
21312-1-AP-150UL
10081-724EA 402 USD
10081-724
Anti-DEPDC6 Rabbit Polyclonal Antibody
Antibodies
DEPTOR, also named as DEPDC6, is a negative regulator of the mTORC1 and mTORC2 signaling pathways. Through mTOR interactions, DEPTOR is implicated in pathways for cell growth and differentiation, as well as cell-cycle prolongation or termination. It inhibits the kinase activity of both complexes. The MW will migrate to 50-55 kDa with phospho- form.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human thyroid cancer Tissue, 1:20-1:200

Type: Primary
Antigen: DEPDC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR