Anti-NHLRC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21310-1-AP-150UL
10091-094EA 402 USD
10091-094
Anti-NHLRC1 Rabbit Polyclonal Antibody
Antibodies
NHLRC1(NHL repeat-containing protein 1) is also named as Malin,EPM2B. It is predicted to encode a 42.3-kDa (395amino acid) protein, malin, containing a detectable zinc-binding RING finger and six NHL-repeat domains.It can suppresses the cellular toxicity of misfolded proteins by promoting their degradation through the ubiquitin-proteasome system (UPS).Defects in NHLRC1 are a cause of progressive myoclonic epilepsy type 2 (EPM2).This antibody is specific to NHLRC1.

Western Blot:L02 Cells, 1:200-1:2000; IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: NHLRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR