Anti-SIX4 Rabbit Polyclonal Antibody

Supplier: Proteintech
21305-1-AP-150UL
10094-662EA 402 USD
10094-662
Anti-SIX4 Rabbit Polyclonal Antibody
Antibodies
The Six (sine oculis) proteins are a family of homeodomain transcription factors that share a conserved DNA-binding domain and are human homologs of the Drosophila sine oculis (so) protein. SIX homeobox 4 (SIX4), also known as AREC3, belongs to the the Six family. Six4 contains one Six domain (which funtions as a homeobox DNA-binding motif) and shares 90% sequence similarity with its mouse counterpart, suggesting that both proteins have similar DNA-binding properties. SIX4 is thought to be involved in neurogenesis, myogenesis, and development of other organs, based on its specific expression in certain neuronal cells of the developing embryo and in adult skeletal muscles.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: SIX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR