Anti-PLK5 Rabbit Polyclonal Antibody

Supplier: Proteintech
21299-1-AP-150UL
10092-478EA 402 USD
10092-478
Anti-PLK5 Rabbit Polyclonal Antibody
Antibodies
PLK5, also named as PLK5P, is an inactive serine/threonine-protein kinase that plays a role in cell cycle progression and neuronal differentiation. This antibody detects the 50-55 kDa (modified form) and ~40 kDa (unmodified form) in human brain tissue lysate.

Western Blot: Mouse Testis Tissue, 1:200-1:1000; IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: PLK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR