Anti-Kv4.2 Rabbit Polyclonal Antibody

Supplier: Proteintech
21298-1-AP-150UL
10089-404EA 537.7 USD
10089-404
Anti-Kv4.2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A549 Cells, 1:500-1:5000

Type: Primary
Antigen: Kv4.2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR